Cytidine-5'-diphosphocholine sodium salt (CDP choline), Hyrdrat
Lieferant: Thermo Scientific
Synonyme:
CDP-Choline, 2-(((((((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)-N,N,N-trimethylethan-1-aminium, sodium salt(1:1), Citicoline sodium salt, CDP-choline sodium salt
CAS No.: 33818-15-4
Molecular Formula: C14H26N4NaO11P2.xH2O
Formula Weight: 510.31(anhy)
Physical Form: Powder
Appearance: White to off-white
Beilstein Reference: 4110040
MDL No.: MFCD00150508
Applications: Intermediate in the synthesis of phosphatidylcholine; also a phospholipase inhibitor
Formel:
C₁₄H₂₆N₄NaO₁₁P₂·xH₂O Molecular Weight: 510,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150508 CAS-Nummer: 33818-15-4 EINECS: 251-689-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...