4-(1-Bromoethyl)benzoic acid 98%
Lieferant: Thermo Scientific
|
Gefahr
|
L13025.03
L13025.06
L13025.03EA
22.9
EUR
L13025.03
L13025.06
4-(1-Bromoethyl)benzoic acid 98%
4-(1-Bromoethyl)benzoic acid
Purity: 98%
CAS No.: 113023-73-7
Molecular Formula: C9H9BrO2
Formula Weight: 229.08
Melting Point: 155-156°
Beilstein Reference: 2084039
UN No.: UN3261
MDL No.: MFCD00020378
Formel:
C₉H₉BrO₂ Molecular Weight: 229,08 g/mol Schmelzpunkt: 155…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020378 CAS-Nummer: 113023-73-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...