Dowex® 50WX4 (H) 100-200 mesh
Lieferant: Thermo Scientific
L13917.18
L13917.30
L13917.0B
L13917.18EA
53.6
EUR
L13917.18
L13917.30
L13917.0B
Dowex® 50WX4 (H) 100-200 mesh
Dowex® 50WX4 (H)
CAS No.: 11113-61-4
MDL No.: MFCD00132725
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00132725 CAS-Nummer: 11113-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...