Agar, körnig NF
Lieferant: Spectrum Chemical
Synonyme:
Agar Agar
SPCMAG105-2.5KGEA
1230
EUR
SPCMAG105-2.5KG
SPCMAG105-50KGBL
Agar, körnig NF
Agar
Form: Granular
Schmelzpunkt:
90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081288 CAS-Nummer: 9002-18-0 EINECS: 232-658-1 Merck Index: 13,00184 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...