Betain, wasserfrei

Lieferant: Spectrum Chemical

Synonyme: TMG

SPCMB1572-25KGBLEA 0 EUR
SPCMB1572-25KGBL
Betain, wasserfrei
Betain
Formel: C₅H₁₁NO₂
Molecular Weight: 117,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012123
CAS-Nummer: 107-43-7
EINECS: 203-490-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR