γ-L-Glutamyl-L-glutamic acid

Lieferant: Thermo Fisher Scientific
ACRO228812500EA 188 EUR
ACRO228812500
γ-L-Glutamyl-L-glutamic acid
Proteine und Peptide
  • Infrared spectrum: White powder
  • Solubility: (5% in water) clear coluorless
  • Specific optical rotation: 4 to 6° (20 °C, 589 nm) (c=1, 0,5 N HCl)
  • Melting point: 179 to 182 °C
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR