Metaldehyd 99%
Lieferant: Thermo Scientific
|
Gefahr
|
ACRO271862500EA
72.9
EUR
ACRO271862500
ACRO271860010
ACRO271860050
Metaldehyd 99%
Metaldehyd
Appearance: White Crystalline powder
Formel:
C₈H₁₆O₄ Molecular Weight: 176,21 g/mol Schmelzpunkt: 246 °C Flammpunkt: 55 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00071549 CAS-Nummer: 108-62-3 UN: 1332 ADR: 4.1,III |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...