Brucine, wasserfrei 99%
Lieferant: Thermo Scientific
|
Gefahr
|
ACRO278720050EA
35
EUR
ACRO278720050
ACRO278721000
ACRO278725000
ACRO278720500
Brucine, wasserfrei 99%
Brucine
Formel:
C₂₃H₂₆N₂O₄ Molecular Weight: 394,47 g/mol Schmelzpunkt: 174…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005942 CAS-Nummer: 357-57-3 UN: 1570 ADR: 6.1,I |
Spezifikation Testergebnisse
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 174 to 179 °C (on dry substance) |
Loss on drying | ≤1 % (105 °C, 3 hrs) |
Sulfated ash | ≤0.1 % |
Specific optical rotation | -118° to -128° (20 °C, 589 nm) (c=2.5, CHCl3) |
Titration with HClO4 | ≥98.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...