Stoddard solvent
Lieferant: Thermo Scientific
|
Gefahr
|
ACRO419730050EA
289
EUR
ACRO419730050
ACRO419730010
Stoddard solvent
Stoddard solvent
Siedepunkt:
149…213 °C (760 mmHg) Schmelzpunkt: –40 °C Dichte: 0,787 g/cm³ Flammpunkt: 40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00132767 CAS-Nummer: 8052-41-3 UN: 1300 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear liquid |
Boiling point | 149 to 213°C (ASTM D 86) |
Density | (15.6°C) 754 to 820 kg/m³ (ASTM D 4052) |
Additional info | copper corrosion: max. 1 (ASTM D 130) |
Additional info | Kauri butanol value: 29 to 45 (ASTM D 1133) |
Color scale | ≤25 APHA (ASTM D 1209) |
Bromine number | ≤5 g Br/100 g (ASTM D 1159) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...