D-δ-Tocopherol 93%

Lieferant: Thermo Scientific

Synonyme: (2R)-2,8-dimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol, (+)-δ-Tocopherol, (R)-2,8-Dimethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, Delta-Tocopherol

421051000 421055000
ACRO421051000EA 197 EUR
ACRO421051000 ACRO421055000
D-δ-Tocopherol 93%
D-delta-Tocopherol

CAS No.: 119-13-1

Formel: C₂₇H₄₆O₂
Molecular Weight: 402,663 g/mol
Flammpunkt: >93 °C
Lagertemperatur: Kühlschrank
MDL: MFCD20486794
CAS-Nummer: 119-13-1

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to brown Viscous liquid
Infrared spectrum Conforms
GC ≥95 %
Color scale ≤25 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR