Acid green 50, rein

Lieferant: Thermo Scientific
Achtung

ACRO190100250EA 48.3 EUR
ACRO190100250
Acid green 50, rein
Acid green 50

Appearance: Dark brown to black Powder

Formel: C₂₇H₂₆N₂O₇S₂
Molecular Weight: 576,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012117
CAS-Nummer: 3087-16-9

Bestellen

Spezifikation Testergebnisse

Appearance Dark brown to black Powder
Infrared spectrum Conforms
UV spectrophotometry ≥50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR