Silica gel, functionalized, Amino-3 ∼1,4 mmol/g, Ø 40-63 micron

Lieferant: Thermo Scientific

ACRO360181000EA 358 EUR
ACRO360181000 ACRO360180250 ACRO360180100
Silica gel, functionalized, Amino-3 ∼1,4 mmol/g, Ø 40-63 micron
Silica gel, functionalized, Amino-3
Formel: C₉H₂₃NO₅Si₂
Molecular Weight: 281,455 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03427643
CAS-Nummer: 71888-97-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Particle size 40 to 63 µm
Loading ≥1.2 mmol/g (molecular)
Nitrogen (N) ≥1.68 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR