Isobutylmagnesium chloride 2 M in Diethylether, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
209521000.EA
63.9
EUR
209521000.
Isobutylmagnesium chloride 2 M in Diethylether, AcroSeal®
Isobutylmagnesium chloride
Appearance: Grey to yellow-brown Solution
Formel:
C₄H₉ClMg Molecular Weight: 116,871 g/mol Dichte: 0,83 g/cm³ Flammpunkt: –040 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000471 CAS-Nummer: 5674-02-2 UN: 3399 ADR: 4.3,I |
Spezifikation Testergebnisse
Appearance | Grey to yellow-brown Solution |
Titration Grignard | 2 M Exact strength on the certificate of analysis |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...