5-Cyan-2-thiophenboronsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO391270010EA
71.2
EUR
ACRO391270010
ACRO391270050
5-Cyan-2-thiophenboronsäure 98%
5-Cyano-2-thiopheneboronic acid
Formel:
C₅H₄BNO₂S Molecular Weight: 152,97 g/mol Schmelzpunkt: 175…180 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02094029 CAS-Nummer: 305832-67-1 UN: 3439 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Beige to pink Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...