3-Benzyloxyphenylhydrazine hydrochloride 98%

Lieferant: Thermo Scientific
Achtung

394020010 394020050
ACRO394020010EA 57 EUR
ACRO394020010 ACRO394020050
3-Benzyloxyphenylhydrazine hydrochloride 98%
3-Benzyloxyphenylhydrazine hydrochloride

Appearance: Violet Powder

Formel: C₁₃H₁₅ClN₂O
Schmelzpunkt: 155…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02258768
CAS-Nummer: 59146-68-8
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Violet Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR