Formaldehyd 4%, gepuffert
Lieferant: KLINIPATH (PL)
Synonyme:
Formol
, Formalin
, Methanal
, Methylaldehyd
VWRK39503-FEA
2.8
EUR
VWRK39503-F
VWRK4187F
VWRK4188-B
VWRK4186
VWRK4177
Formaldehyd 4%, gepuffert
Formaldehyd
Formaldehyde solution 4% (pH = 7).
- Pre-filled solutions
For use as a general purpose histological fixative.
Verpackung: In ready-to-use screw cap PTFE or glass containers.
Formel:
CH₂O Molecular Weight: 30,03 g/mol |
MDL:
MFCD00003274 CAS-Nummer: 50-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...