3-Methylisochinolin 98%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO428440010EA
63.9
EUR
ACRO428440010
ACRO428440050
3-Methylisochinolin 98%
3-Methylisoquinoline
Formel:
C₁₀H₉N Molecular Weight: 143,19 g/mol Siedepunkt: 251 °C (760 mmHg) Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024139 CAS-Nummer: 1125-80-0 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to yellow to brown Crystalline powder or solid |
Infrared spectrum | Authentic |
Melting point | 60 to 67 °C |
HPLC | ≥97.5 % |
Density | (15 °C) ≥0.79 kg/dm³ |
Aromatics | ≤0.2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...