1-Chlorbutan 99.5% zur Analyse

Lieferant: Thermo Scientific
Gefahr

Synonyme: Butylchlorid

ACRO447020010EA 72.9 EUR
ACRO447020010
1-Chlorbutan 99.5% zur Analyse
1-Chlorbutan
Formel: CH₃(CH₂)₃Cl
Molecular Weight: 92,57 g/mol
Siedepunkt: 77…78 °C (760 mmHg)
Schmelzpunkt: –123 °C
Dichte: 0,88 g/cm³
Flammpunkt: –12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001009
CAS-Nummer: 109-69-3
UN: 1127
ADR: 3,II
https://de.vwr.com/stibo/web/std.lang.all/58/18/725818.jpg

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4030 (20°C, 589 nm)
Water ≤0.05 %
GC ≥99.4 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity ≤0.0005 meq/g
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Sulfur compounds (as S) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR