Ciprofloxacin 98%

Lieferant: Thermo Scientific

Synonyme: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid

449620050 449620250
ACRO449620050EA 32.1 EUR
ACRO449620050 ACRO449620250
Ciprofloxacin 98%
Ciprofloxacin
Formel: C₁₇H₁₈FN₃O₃
Molecular Weight: 331,35 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00185755
CAS-Nummer: 85721-33-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Sulfated ash ≤0.1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR