Decamethylcyclopentasiloxan 97%
Lieferant: Thermo Scientific
Synonyme:
Siliconöl DC 245, Volasil 245, Cyclopentasiloxan, Dow Corning 245® fluid
454580500
454582500
ACRO454580500EA
32.3
EUR
ACRO454580500
ACRO454582500
Decamethylcyclopentasiloxan 97%
Decamethylcyclopentasiloxan
Formel:
C₁₀H₃₀O₅Si₅ Molecular Weight: 370,77 g/mol Siedepunkt: 101 °C (20 mmHg) Schmelzpunkt: -44 °C Dichte: 0,959 g/cm³ Flammpunkt: 76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046966 CAS-Nummer: 541-02-6 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3970 to 1.3990 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...