2-Pyridinsulfonsäure 97%

Lieferant: Thermo Scientific
Gefahr

ACRO455760050EA 86.7 EUR
ACRO455760050
2-Pyridinsulfonsäure 97%
2-Pyridinesulphonic acid
Formel: C₅H₅NO₃S
Molecular Weight: 159,1631 g/mol
Schmelzpunkt: 244…249 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234080
CAS-Nummer: 15103-48-7
UN: 2585
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 245 to 252 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR