Bunded spill flooring, PE
Lieferant: asecos
MSGA8828EA 1710.5 EUR
MSGA8828 MSGA31396 MSGA31397 MSGA8829 MSGA19234 MSGA31394 MSGA31395 MSGA8827 MSGA19236 MSGA19235 MSGA17301 MSGA8684
Bunded spill flooring, PE
Absorptionsmaterial
The bunded spill flooring made from PE is ideal for covering a large storage area.
  • Compact and robust design – safe and approved
  • Approved for the safe storage of water pollutants
  • Available with galvanised or polyethylene grid
  • High resistance against chemicals
  • Wheel load 150 kg
Load capacity: 800 kg/m²