siRNA transfection reagent, TransIT-siQUEST®
Lieferant: MIRUS
MIR2110
MIR2116
MIR2115
MIR2114
MIRUMIR2110EA
794
EUR
MIRUMIR2110
MIRUMIR2116
MIRUMIR2115
MIRUMIR2114
siRNA transfection reagent, TransIT-siQUEST®
Transfektionsreagenzien
TransIT-siQUEST® is a broad spectrum siRNA transfection reagent that enables high efficiency siRNA delivery and knockdown of target gene expression in many cell types including primary cells.
- Broad spectrum siRNA delivery which utilises one transfection reagent and protocol for a wide variety of cells
- Low cellular toxicity that maintain cell density and reduce experimental biases
- Reproducible results providing routinely consistent, targeted gene knockdowns
- High knockdown efficiency ensures optimal gene silencing in a large percentage of cells
Transfections with TransIT-siQUEST do not require medium changes and can be carried out in serum-containing medium.