Plasmid delivery controls, Label IT®
Lieferant: MIRUS
MIR7907 MIR7906 MIR7905 MIR7904
MIRUMIR7907EA 967 EUR
MIRUMIR7907 MIRUMIR7906 MIRUMIR7905 MIRUMIR7904
Plasmid delivery controls, Label IT®
Vektoren, Plasmide und Bibliotheken
Fluorescently labelled plasmids ideal for in vitro and in vivo tracking.
  • For in vivo delivery
  • Easily visualise transfected cells and assess delivery efficiency using fluorescent microscopy
  • Suitable for co-delivery experiments with functional plasmids
Lieferumfang: Supplied as a ready to use 0,5 mg/ml concentrate.