Beef extract, tan powder
Lieferant: MP Biomedicals
ICNA0210088390EA 360 EUR
ICNA0210088390 ICNA0210088380 ICNA0210088350
Beef extract, tan powder
Medien für die Mikrobiologie Basismaterialien für die Mikrobiologie
Beef extract is a water soluble powder, standardized for use in microbiological media.
  • Soluble in water
Beef extract is used in general purpose media. It is recommended for use in bacteriological examination of water, milk, and other materials, where uniform composition of media is important and for biochemical studies, particularly fermentation reactions because of its independence from fermentable substances.