γ-(N-Morpholino)propansulfonsäure (MOPS), weißes Pulver
Lieferant: MP Biomedicals
Synonyme: 3-(N-Morpholino)propansulfonsäure
0210237010 0210237080 0210237083 0210237091
ICNA0210237010EA 35.4 EUR
ICNA0210237010 ICNA0210237080 ICNA0210237083 ICNA0210237091
γ-(N-Morpholino)propansulfonsäure (MOPS), weißes Pulver
γ-(N-Morpholino)propansulfonsäure (MOPS)
MOPS is a zwitterionic amino acid which acts as one of the "Good" buffers.
MOPS is used as a biological buffer.
Concentrations higher than 20 mM should not be used with mammalian cell cultures.
Soluble in water (to approximately 33% w/w - clear, colorless solution)
Bitte beachten Research Use Only (RUO).
Formel: C₇H₁₅NO₄S
Molecular Weight: 209,27 g/mol
Schmelzpunkt: 277 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006183
CAS-Nummer: 1132-61-2
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard
UV/Vis: (1M aq soln) OD250 nm ≤0.04
UV/Vis: (1M aq soln) OD280 nm ≤0.04
pH (10% aq soln) 3.2 - 4.2