Toluidinblau O, Pulver C.I. 52040
Lieferant: MP Biomedicals
Synonyme: Toloniumchlorid3-Amino-7-(dimethylamino)-2-methyl-5-phenothiaziniumchlorid
ICNA0215264925EA 114 EUR
ICNA0215264925
Toluidinblau O, Pulver C.I. 52040
Toluidinblau O
Toluidine blue O is useful as a metachromatic nuclear stain.
Soluble in water (10 mg/ml, blue violet solution), ethanol (4 mg/ml), alcohol (blue solution).
Formel: C₁₅H₁₆ClN₃S
Molecular Weight: 305,83 g/mol
MDL: MFCD00011934
CAS-Nummer: 92-31-9
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
UV/VIS: λmax (H₂O) 630 ± 4 nm