ICNA02157451
Lieferant: MP Biomedicals
Synonyme:
2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthincarbonsäure
, 2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalin-3-carbonsäure
, 3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalin-2-carbonsäure
, Cal-Red®
ICNA0215745105EA
48.2
EUR
ICNA0215745105
ICNA02157451
Patton-Reeder-Reagenz (Calconcarbonsäure)
2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acid is a useful indicator for calcium titrations with EDTA in the presence of magnesium.
Soluble in methanol (clear reddish violet solution), 50 mg plus 5 ml of 1 M NaOH; slightly soluble in water.
Formel:
C₂₁H₁₄N₂O₇S Molecular Weight: 438,42 g/mol |
MDL:
MFCD00004078 CAS-Nummer: 3737-95-9 |
Spezifikation Testergebnisse
Identity Test | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...