Kifunensine ≥98%, weißes Pulver

Lieferant: MP Biomedicals

ICNA0215995201EA 187 EUR
ICNA0215995201
Kifunensine ≥98%, weißes Pulver
Kifunensine

Kifunensine is an alkaloid compound, originally isolated from the actinomycete Kitasporia kifunensis, that exhibits potent and selective inhibition of class I α-mannosidases. Kifunensine is used to suppress Endoplasmic Reticulum-Associated Degradation (ERAD) via the inhibition of endoplasmic reticulum-associated mannosidase activity.


Selective inhibitor of class I glycoprotein-processing α-mannosidases.


Soluble in water (25 mg/ml).

Formel: C₈H₁₂N₂O₆
Molecular Weight: 232,192 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 109944-15-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR