Scalpels, disposable, Feather®
Lieferant: PFM Medical
FEAT200130023EA 23 EUR
FEAT200130023 FEAT200130022 FEAT200130011 FEAT200130021 FEAT200130010 FEAT200130020 FEAT02.00210015 FEAT200130015 FEAT200130025 FEAT200130024
Scalpels, disposable, Feather®
Skalpelle Skalpelle, komplett
These disposable standard scalpels with high-quality stainless steel blade are suitable for all surgical applications.
  • Outstanding quality and performance thanks to optimised micrometer-accurate precision grinding
  • Double-honing process ensures ultra-sharp cutting edges of great durability
Verpackung: 20 pieces per pack.