Ball valve, Parker Balston® shut-off gas valve
Lieferant: Restek
REST22193EA 172 EUR
REST22193 REST22192 REST22195 REST22194
Ball valve, Parker Balston® shut-off gas valve
Ventile Vakuum-Ventile
Ball valves have a floating ball to assist sealing and to reduce operating torque, and dual seats to provide leak-tight bidirectional sealing. They are rated to 1500 psig at 21 °C and have a maximum temperature of about 177 °C.