Pierce™ Background Eliminator
Lieferant: Thermo Fisher Scientific
Pierce™
PIER21065EA 212 EUR
PIER21065
Pierce™ Background Eliminator
Filmbearbeitungs-Materialien
High background, shading, overexposed bands and speckling are problems inherent to film exposure. The kit can correct these problems without the need to re-expose the blot to film or re-do the experiment. Pierce™ Background Eliminator solution can be used with newly exposed films or exposed films that have been stored. It can also be used with any brand of film.
  • Fast, easy background elimination from film
  • Eliminates unwanted background from overexposed, speckled or shaded films
  • No need for time-consuming re-exposures to find the optimal image
  • No need to re-optimise assay reagents in order to obtain the optimal image