Plastic refrigerator/freezer thermometer
Lieferant: TFA Dostmann
620-8204EA 3.9 EUR
620-8204
Plastic refrigerator/freezer thermometer
Thermometer Analoge Thermometer
This useful plastic thermometer shows you at a glance whether vegetables, cheese, meat, etc. are properly cooled.
  • You can easily hang in your fridge or freezer with the handy mounting part
  • Temperature is measured with proven bimetallic technology
  • With freezing-zones indication for easy reading
The product is entirely manufactured in Germany.