It binds to protein kinase C and leads to a very strong activation of this enzyme. Physiological effects are very diverse.
TPA is soluble in DMSO or ethanol. The crystalline substance as well as the stock solution should be stored in the dark at −20 °C (stock solution 1 mg/ml; working concentration 100 ng/ml). Long term storage should be at −70 °C.