Dropless pipette
Lieferant: COPAN Diagnostics
COPA214CEA 743 EUR
COPA214C
Dropless pipette
Pipetten Pasteur- und Transfer-Pipetten
LDPE, disposable.
  • No drop counting; just fill the pipette and release
  • Delivers accurate volume in one step
  • Simplifies rapid membrane test protocol
  • Ideal for OTC kits
  • Built-in procedural control for in vitro diagnostic use e.g. home pregnancy test kits