2-Mercaptoethansulfonsäure Natriumsalz (Mesna) 98% analytischer Standard
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Sodium 2-mercaptoethanesulphonate
443150050
443150100
443150250
ACRO443150050EA
49.9
EUR
ACRO443150050
ACRO443150100
ACRO443150250
2-Mercaptoethansulfonsäure Natriumsalz (Mesna) 98% analytischer Standard
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
Formel:
C₂H₅NaO₃S₂ Molecular Weight: 164,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007535 CAS-Nummer: 19767-45-4 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration Iodometric | ≥97.5 to 102 % (On dried substance) |
Loss on drying | ≤2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...