Citicoline 98%

Lieferant: Thermo Scientific

Synonyme: Cytidine 5'-diphosphocholine, 2-(((((((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)-N,N,N-trimethylethanaminium

457780010 457780050 457780250
ACRO457780010EA 49.2 EUR
ACRO457780010 ACRO457780050 ACRO457780250
Citicoline 98%
Citicoline
Formel: C₁₄H₂₆N₄O₁₁P₂
Molecular Weight: 488,33 g/mol
Schmelzpunkt: 240…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00868097
CAS-Nummer: 987-78-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR