Cefixime 98%

Lieferant: Thermo Scientific

Synonyme: (6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-((carboxymethoxy)imino)acetamido)-8-oxo-3-vinyl-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

459480010 459480050
ACRO459480010EA 33.9 EUR
ACRO459480010 ACRO459480050
Cefixime 98%
Cefixime
Formel: C₁₆H₁₅N₅O₇S₂
Molecular Weight: 453,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00865020
CAS-Nummer: 79350-37-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige to light brown Powder
Infrared spectrum Conforms
Water 9.0 to 12.0 %
HPLC ≥97.5 %
pH 2.6 to 4.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR