O-Diphenylphosphinylhydroxylamine 96%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO461850010EA
308
EUR
ACRO461850010
ACRO461850050
O-Diphenylphosphinylhydroxylamine 96%
O-Diphenylphosphinylhydroxylamine
Formel:
C₁₂H₁₂NO₂P Schmelzpunkt: 104 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD12755701 CAS-Nummer: 72804-96-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...