n-Decan ≥95%, Sigma-Aldrich®

Lieferant: Merck
Gefahr

SIAL30570-1LEA 91.4 EUR
SIAL30570-1L SIAL30570-2.5L
n-Decan ≥95%, Sigma-Aldrich®
n-Decan

Decane was used as solvent to investigate the self-association of cyclohexanols.


Decane is an organic solvent. Bifunctional conversion of decane over Pt/ZSM-22, Pt/ZSM-5 and Pt/USY catalysts was compared. Structure of single-phase (sodium di-2-ethylsulfosuccinate)/D2O/decane microemulsions was investigated by small-angle neutron scattering.

Formel: H₃C(CH₂)₈CH₃
Molecular Weight: 142,28 g/mol
Siedepunkt: 170…182 °C
Schmelzpunkt: –30 °C (Lit.)
Dichte: 0,730 g/ml (20 °C)
Flammpunkt: 46 °C
MDL: MFCD00008954
CAS-Nummer: 124-18-5
https://de.vwr.com/stibo/web/std.lang.all/60/32/736032.jpg

Bestellen

Spezifikation Testergebnisse

Assay ≥95%
Vapor density (vs air) 4,9
Vapor pressure (16,5 °C) 1 mmHg
Vapor pressure ( 37,7 °C) 3,77 mmHg
Autoignition temperature 210 °C
Explosion limit 2,6%
Refractive index n20/D 1,412
Density (20 °C) 0,730 g/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR