Gold ≥99,999% (Metall-Basis), Drahtform, Premion®, Ø 1.0 mm (0.04 in)

Lieferant: Thermo Scientific

Synonyme: Aurum

14726.BSEA 418 EUR
14726.BS 14726.BP 14726.BW
Gold ≥99,999% (Metall-Basis), Drahtform, Premion®, Ø 1.0 mm (0.04 in)
Gold
Formel: Au
Molecular Weight: 196,97 g/mol
Siedepunkt: 2800 °C (1013 hPa)
Schmelzpunkt: 1063 °C
Dichte: 19,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003436
CAS-Nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR