4'-Chlorchalkon
Lieferant: FLUOROCHEM LTD (CHEMPUR)
Synonyme:
4'-Chlorbenzylidenacetophenon
FLUO011129-25GEA
0
EUR
FLUO011129-25G
FLUO011129-100G
4'-Chlorchalkon
4'-Chlorchalkon
Formel:
C₁₅H₁₁ClO Molecular Weight: 242,7 g/mol |
MDL:
MFCD00016343 CAS-Nummer: 956-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...