4'-Chlorchalkon

Lieferant: MENAI (PART OF CHEMPUR)

Synonyme: 4'-Chlorbenzylidenacetophenon

MENAIP24.250EA 0 EUR
MENAIP24.250 MENAIP24.100
4'-Chlorchalkon
4'-Chlorchalkon
Formel: C₁₅H₁₁ClO
Molecular Weight: 242,7 g/mol
MDL: MFCD00016343
CAS-Nummer: 956-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR