4'-Fluorchalkon

Lieferant: FLUOROCHEM

Synonyme: 4'-Fluorbenzylidenacetophenon

005222-25G 005222-5G
FLUO005222-25GEA 317 EUR
FLUO005222-25G FLUO005222-5G
4'-Fluorchalkon
4'-Fluorchalkon
Formel: C₁₅H₁₁FO
Molecular Weight: 226,25 g/mol
MDL: MFCD00017961
CAS-Nummer: 399-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR