Bienenwachs, weiß, Pastillenform, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®

Lieferant: Merck

1.11544.9020EA 0 EUR
1.11544.9020
Bienenwachs, weiß, Pastillenform, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Beeswax
Schmelzpunkt: ∼64 °C
Dichte: 0,95 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00132754
CAS-Nummer: 8012-89-3
EINECS: 232-383-7

Bestellen

Spezifikation Testergebnisse

Appearance white or yellowish-white pastillic pieces
Dropping point 61 - 66 °C
Acid value 17.0 - 24.0
Saponification value 87 - 104
Ester value 70 - 80
Ceresin,paraffins and other waxes conforms
Glycerol and other polyols ≤ 0.5 %
Residual solvents (Ph.Eur./ICH) excluded by production process

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR