Anthracen-[D10] (98% D)

Lieferant: LGC Standards PROMOCHEM

Synonyme: Paranaphthalin-D10

PROOCIL-DLM-102-1EA 243 EUR
CAMBDLM-102-1 PROOCIL-DLM-102-1 PROOCIL-DLM-102-5
Anthracen-[D10] (98% D)
Anthracen-[D10]
Formel: C₁₄D₁₀
Molecular Weight: 188,15 g/mol
Siedepunkt: 340 °C (1013 hPa)
Lagertemperatur: Kühlschrank
CAS-Nummer: 1719-06-8
EINECS: 217-004-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR