D(+)-Maltose Monohydrat
Lieferant: Merck
Synonyme:
Malzzucker
47288.EA
41.6
EUR
47288.
D(+)-Maltose Monohydrat
D(+)-Maltose Monohydrat
Formel:
C₁₂H₂₂O₁₁·1H₂O Molecular Weight: 360,32 g/mol Schmelzpunkt: 160…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149343 CAS-Nummer: 6363-53-7 EINECS: 200-716-5 Merck Index: 13,05736 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...