D(+)-Trehalose, wasserfrei 99%
Lieferant: Thermo Scientific
Synonyme:
(+)-Trehalose, α,α-Trehalose, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol), Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside, D-(+)-Trehalose Anhydrous, D-Trehalose
309870250
309871000
309875000
ACRO309870250EA
118
EUR
ACRO309870250
ACRO309871000
ACRO309875000
D(+)-Trehalose, wasserfrei 99%
D(+)-Trehalose
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342,3 g/mol Schmelzpunkt: 203 °C Dichte: 1,53 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006628 CAS-Nummer: 99-20-7 EINECS: 202-739-6 Merck Index: 12,09713 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1.0 % (K.F.) |
Sulfated ash | ≤0.1 % (on dry basis) |
Solubility | (10 % in water) Clear colorless to light yellow |
HPLC | ≥98.5 % (ELSD) on dry basis |
Specific optical rotation | +194° to +200° (20°C, 589 nm) (c=7, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...