D(-)-Arabinose 99+%

Lieferant: Thermo Scientific

ACRO161451000EA 169 EUR
ACRO161451000 ACRO161450250
D(-)-Arabinose 99+%
D(-)-Arabinose
Formel: C₅H₁₀O₅
Molecular Weight: 150,13 g/mol
Schmelzpunkt: 152…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135608
CAS-Nummer: 28697-53-2
EINECS: 233-708-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.1 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -101.0° to 106.0° (20°C, 589 nm) (c=4, H2O) (after 24 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR