D(-)-Arabinose ≥99%

Lieferant: Thermo Scientific

A10357.PGEA 0 EUR
A10357.PG A10357.18 A10357.09 A10357.30
D(-)-Arabinose ≥99%
D(-)-Arabinose
Formel: C₅H₁₀O₅
Molecular Weight: 150,13 g/mol
Schmelzpunkt: 154…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135608
CAS-Nummer: 10323-20-3
EINECS: 233-708-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR